Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039521.1 | NP_001034610.1 | 6 | 557 | |
GenBank mRNA | AK080397.1 | - | 6 | 512 | |
AK080680.1 | - | 538 | |||
AK165676.1 | - | 6 | 546 | ||
BC034110.1 | - | 2 | 72 | ||
BC138774.1 | - | 6 | 522 | 8861796 | |
BC138776.1 | - | 6 | 522 | 8861798 |