Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134155.1 | NP_598916.1 | 4 | 343 | |
GenBank mRNA | AF233580.1 | - | 4 | 354 | |
BC003485.1 | - | 4 | 361 | 2811430 | |
BC016108.1 | - | 4 | 343 | 4921658 | |
BC038380.1 | - | 2582 |