Gene Transcripts

Gene Symbol
Kif24
Entrez Gene ID
109242
Gene Name
kinesin family member 24
Gene Aliases
4933425J19Rik, 9430029L23Rik
Chromosome Location
Chr. 4: 41390748 - 41464933 on Build GRCm38
UniGene ID
Mm.370288
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024241.2 NP_077203.2 6, 7 1359
GenBank mRNA AK050683.1 - 6, 7 1345
AK135307.1 - 6, 7 1360
BC067395.1 - 6, 7 1351 30460089

View Full Product Details