Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009777.2 | NP_033907.1 | 2 | 192 | |
GenBank mRNA | AK140712.1 | - | 2 | 149 | |
AK152764.1 | - | 2 | 211 | ||
AK153124.1 | - | 2 | 84 | ||
BC067001.1 | - | 2 | 128 | 5715633 | |
M22531.1 | - | 2 | 192 | ||
M36293.1 | - | 2 | 192 |