Gene Transcripts

Gene Symbol
Cbr2
Entrez Gene ID
12409
Gene Name
carbonyl reductase 2
Gene Aliases
MLCR
Chromosome Location
Chr. 11: 120729485 - 120732025 on Build GRCm38
UniGene ID
Mm.21454
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007621.2 NP_031647.1 5 477
GenBank mRNA AK081816.1 - 5 448
BC010758.1 - 5 437 4219349
D26123.1 - 5 464
X07411.1 - 5 434

View Full Product Details