Gene Transcripts
- Gene Symbol
- Ccnd1
- Entrez Gene ID
- 12443
- Gene Name
- cyclin D1
- Gene Aliases
- AI327039, CycD1, Cyl-1, PRAD1, bcl-1, cD1
- Chromosome Location
- Chr. 7: 144929931 - 144939937 on Build GRCm38
- UniGene ID
- Mm.273049
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007631.2 |
NP_031657.1
|
3, 4 |
810 |
|
GenBank mRNA |
AK005352.1 |
- |
3, 4 |
504 |
|
AK017834.1 |
- |
3, 4 |
774 |
|
AK053348.1 |
- |
3, 4 |
718 |
|
AK149905.1 |
- |
3, 4 |
717 |
|
AK164997.1 |
- |
3, 4 |
755 |
|
AK170833.1 |
- |
3, 4 |
691 |
|
BC044841.1 |
- |
3, 4 |
713 |
3155470 |
BC060172.1 |
- |
3, 4 |
705 |
|
EU600170.1 |
- |
3, 4 |
810 |
|
M64403.1 |
- |
3, 4 |
765 |
|
S78355.1 |
- |
3, 4 |
715 |
|
View Full Product Details