Gene Transcripts

Gene Symbol
Cdk5r2
Entrez Gene ID
12570
Gene Name
cyclin-dependent kinase 5, regulatory subunit 2 (p39)
Gene Aliases
B230310J22Rik, p39
Chromosome Location
Chr. 1: 74855029 - 74857732 on Build GRCm38
UniGene ID
Mm.153833
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009872.3 NP_034002.2 1 375
GenBank mRNA BC103550.1 - 1 303

View Full Product Details