Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009894.3 | NP_034024.2 | 1, 2 | 175 | |
GenBank mRNA | AF041377.1 | - | 1, 2 | 163 | |
AK003968.1 | - | 1, 2 | 212 | ||
BC012664.1 | - | 1, 2 | 139 | 4218866 | |
BC046340.1 | - | 1, 2 | 169 | 6478230 | |
CB600615.1 | - | 1, 2 | 152 |