Gene Transcripts

Gene Symbol
Crybb1
Entrez Gene ID
12960
Gene Name
crystallin, beta B1
Gene Aliases
3110006K12Rik, BB1CRY
Chromosome Location
Chr. 5: 112254477 - 112269582 on Build GRCm38
UniGene ID
Mm.29488
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023695.2 NP_076184.1 5 524
GenBank mRNA AF106853.1 - 5 524
AK014012.1 - 5 526
AK053869.1 - 5 525
BC029012.1 - 4 508 5358714

View Full Product Details