Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001276461.1 | NP_001263390.1 | 3 | 382 | |
NM_001276462.1 | NP_001263391.1 | 1 | 291 | ||
NM_001276463.1 | NP_001263392.1 | 1 | 291 | ||
NM_001276467.1 | NP_001263396.1 | 3 | 382 | ||
NM_010026.3 | NP_034156.2 | 3 | 382 | ||
GenBank mRNA | AF075461.1 | - | 3 | 201 | |
AF075462.1 | - | 3 | 201 | ||
AK132979.1 | - | 1 | 121 | ||
AK146545.1 | - | 3 | 220 | ||
AK147689.1 | - | 1 | 292 | ||
AK161586.1 | - | 292 | |||
BC048818.1 | - | 1 | 257 | ||
BC094581.1 | - | 3 | 382 | 30549241 | |
CB527210.1 | - | 1 | 85 | ||
CF735731.1 | - | 3 | 382 |