Gene Transcripts

Gene Symbol
Adam28
Entrez Gene ID
13522
Gene Name
a disintegrin and metallopeptidase domain 28
Gene Aliases
C130072N01Rik, D430033C21Rik, Dtgn1, MDC-L, MDC-Lm, MDC-Ls, MDCL, TECADAM, eMDCII
Chromosome Location
Chr. 14: 68604994 - 68655887 on Build GRCm38
UniGene ID
Mm.117450
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048175.2 NP_001041640.1 13, 14 1523
NM_010082.2 NP_034212.1 13, 14 1523
NM_183366.3 NP_899222.1 13, 14 1523
GenBank mRNA AF153350.1 - 13, 14 1430
AF163290.1 - 13, 14 1523
AF163291.1 - 13, 14 1523
AF163292.1 - 13, 14 1523
AF163293.1 - 13, 14 1523
AK136715.1 - 4, 5 427
BC058782.1 - 13, 14 1398 6310763
U06146.1 - 1, 2 90

View Full Product Details