Gene Transcripts
- Gene Symbol
- Fbln1
- Entrez Gene ID
- 14114
- Gene Name
- fibulin 1
- Gene Aliases
- Chromosome Location
- Chr. 15: 85205944 - 85286294 on Build GRCm38
- UniGene ID
- Mm.297992
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_010180.2 |
NP_034310.2
|
7, 8 |
906 |
|
GenBank mRNA |
AK035388.1 |
- |
7, 8 |
859 |
|
AK083573.1 |
- |
7, 8 |
909 |
|
AK085989.1 |
- |
7, 8 |
849 |
|
AK086451.1 |
- |
7, 8 |
907 |
|
AK137511.1 |
- |
7, 8 |
849 |
|
AK159646.1 |
- |
7, 8 |
861 |
|
AK211445.1 |
- |
1, 2 |
45 |
|
AK219641.1 |
- |
1, 2 |
45 |
|
BC007140.1 |
- |
7, 8 |
850 |
3495754 |
EU543224.1 |
- |
7, 8 |
844 |
|
X70854.1 |
- |
7, 8 |
903 |
|
View Full Product Details