Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001077189.1 | NP_001070657.1 | 8 | 1061 | |
NM_010187.2 | NP_034317.1 | 7 | 920 | ||
GenBank mRNA | AK089266.1 | - | 7 | 921 | |
AK150613.1 | - | 7 | 921 | ||
AK155329.1 | - | 8 | 1053 | ||
AK170035.1 | - | 7 | 910 | ||
AK171254.1 | - | 8 | 1053 | ||
AK171316.1 | - | 8 | 1044 | ||
BC011305.1 | - | 7 | 883 | ||
BC019758.1 | - | 7 | 883 | ||
BC038070.1 | - | 7 | 907 | 3993915 | |
CD773836.1 | - | 1 | 492 | ||
M14216.1 | - | 7 | 878 | ||
M16367.1 | - | 10 | 1299 | ||
M17515.1 | - | 8 | 1015 | ||
U31801.1 | - | 6 | 876 | ||
U31802.1 | - | 6 | 876 | ||
U31803.1 | - | 5 | 735 | ||
U31804.1 | - | 5 | 735 | ||
U51629.1 | - | 8 | 885 |