Gene Transcripts

Gene Symbol
Fhl3
Entrez Gene ID
14201
Gene Name
four and a half LIM domains 3
Gene Aliases
SLIM2
Chromosome Location
Chr. 4: 124695963 - 124708611 on Build GRCm38
UniGene ID
Mm.331856
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010213.3 NP_034343.2 5 694
GenBank mRNA AF114382.1 - 5 665
AF149826.1 - 4 492
BC145939.1 - 5 658 40131242
CB205184.1 - 3 351

View Full Product Details