Gene Transcripts
- Gene Symbol
- Gfra1
- Entrez Gene ID
- 14585
- Gene Name
- glial cell line derived neurotrophic factor family receptor alpha 1
- Gene Aliases
- AU042498
- Chromosome Location
- Chr. 19: 58235581 - 58455946 on Build GRCm38
- UniGene ID
- Mm.88367
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001285457.1 |
NP_001272386.1
|
2, 3 |
570 |
|
NM_010279.3 |
NP_034409.1
|
3, 4 |
771 |
|
GenBank mRNA |
AB000800.1 |
- |
2, 3 |
325 |
|
AF015172.1 |
- |
2, 3 |
553 |
|
AK076171.1 |
- |
3, 4 |
691 |
|
BC011532.1 |
- |
2, 3 |
516 |
|
BC040251.1 |
- |
2, 3 |
354 |
|
BC044783.2 |
- |
3, 4 |
650 |
|
BC054378.1 |
- |
2, 3 |
442 |
6390018 |
KM079499.1 |
- |
2, 3 |
430 |
|
View Full Product Details