Gene Transcripts

Gene Symbol
Tsc22d3
Entrez Gene ID
14605
Gene Name
TSC22 domain family, member 3
Gene Aliases
DIP, Dsip1, Gilz, TSC-22R, Tilz3
Chromosome Location
Chr. X: 140539528 - 140600522 on Build GRCm38
UniGene ID
Mm.22216
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077364.1 NP_001070832.1 3, 4 578
NM_010286.4 NP_034416.3 2, 3 403
GenBank mRNA AF024519.1 - 2, 3 388
AF201287.1 - 2, 3 147
AF201288.1 - 2, 3 383
AF201289.1 - 3, 4 540
AK083389.1 - 2, 3 402
AK144196.1 - 2, 3 406
BC028813.1 - 2, 3 393 5136440
BY071907.1 - 2, 3 220
EU818782.1 - 3, 4 577

View Full Product Details