Gene Transcripts

Gene Symbol
Gucy2e
Entrez Gene ID
14919
Gene Name
guanylate cyclase 2e
Gene Aliases
GC-E, GC1, ROS-GC1
Chromosome Location
Chr. 11: 69218117 - 69237358 on Build GRCm38
UniGene ID
Mm.291816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008192.3 NP_032218.2 4 1366
GenBank mRNA AK044282.1 - 4 1367
BC057954.1 - 4 1351

View Full Product Details