Gene Transcripts

Gene Symbol
Foxg1
Entrez Gene ID
15228
Gene Name
forkhead box G1
Gene Aliases
2900064B05Rik, BF-1, Bf1, Hfh9, Hfhbf1
Chromosome Location
Chr. 12: 49382883 - 49386867 on Build GRCm38
UniGene ID
Mm.4704
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001160112.1 NP_001153584.1 1 2998
NM_008241.2 NP_032267.1 2 1960
GenBank mRNA AK132305.1 - 1 2986
BC046958.1 - 1 1598 6314329
BC055774.1 - 1 1292
BC064449.1 - 1 1903
BC079597.1 - 1 2664
U36760.1 - 2 1960

View Full Product Details