Gene Transcripts
- Gene Symbol
- Hsd17b4
- Entrez Gene ID
- 15488
- Gene Name
- hydroxysteroid (17-beta) dehydrogenase 4
- Gene Aliases
- 17-beta-HSD, 17[b]-HSD, 4, DBP, MFE-2, MFP2, MPF-2, Mfp-2, perMFE-2
- Chromosome Location
- Chr. 18: 50128201 - 50196270 on Build GRCm38
- UniGene ID
- Mm.277857
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_008292.4 |
NP_032318.2
|
20, 21 |
1922 |
|
GenBank mRNA |
AK004866.1 |
- |
20, 21 |
1922 |
|
AK088381.1 |
- |
20, 21 |
1897 |
|
AK147182.1 |
- |
20, 21 |
1894 |
|
AK152532.1 |
- |
20, 21 |
1816 |
|
AK161655.1 |
- |
20, 21 |
1817 |
|
AK166801.1 |
- |
20, 21 |
1835 |
|
AK167060.1 |
- |
20, 21 |
1872 |
|
AK169077.1 |
- |
20, 21 |
1824 |
|
AK208618.1 |
- |
2, 3 |
203 |
|
AK218815.1 |
- |
2, 3 |
203 |
|
BC022175.1 |
- |
20, 21 |
1795 |
4209394 |
X89998.1 |
- |
20, 21 |
1783 |
|
View Full Product Details