Gene Transcripts

Gene Symbol
Ier5
Entrez Gene ID
15939
Gene Name
immediate early response 5
Gene Aliases
Chromosome Location
Chr. 1: 155096367 - 155099636 on Build GRCm38
UniGene ID
Mm.12246
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010500.2 NP_034630.1 1 978
GenBank mRNA AF079527.1 - 1 978
AK146609.1 - 1 975
AK150461.1 - 1 977
BC051986.1 - 1 952 5707524

View Full Product Details