Gene Transcripts

Gene Symbol
Kif7
Entrez Gene ID
16576
Gene Name
kinesin family member 7
Gene Aliases
Chromosome Location
Chr. 7: 79698098 - 79715772 on Build GRCm38
UniGene ID
Mm.101633
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291222.1 NP_001278151.1 11 2521
NM_010626.3 NP_034756.2 11 2521
GenBank mRNA BC034093.1 - 1 93
BC141045.1 - 11 2522 9055657
BC145222.1 - 11 2522 9053773

View Full Product Details