Gene Transcripts

Gene Symbol
Fabp5
Entrez Gene ID
16592
Gene Name
fatty acid binding protein 5, epidermal
Gene Aliases
E-FABP, Fabpe, Klbp, PA-FABP, mal1
Chromosome Location
Chr. 3: 10012585 - 10016610 on Build GRCm38
UniGene ID
Mm.741
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001272097.1 NP_001259026.1 1 85
NM_001272098.1 NP_001259027.1 1 85
NM_010634.3 NP_034764.1 1 85
GenBank mRNA AK008782.1 - 1 79
AK011551.1 - 1 81
AK166506.1 - 1 80
AK167389.1 - 1 80
BC002008.1 - 1 65 3490535
BC053042.1 - 1 65
BC100543.1 - 1 60 30136900
W54445.1 - 1 38
X70100.1 - 1 64

View Full Product Details