Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023377.4 | NP_075866.2 | 3 | 272 | |
GenBank mRNA | AF480302.1 | - | 3 | 248 | |
AK009980.1 | - | 3 | 278 | ||
AK077328.1 | - | 3 | 290 | ||
AK080060.1 | - | 3 | 290 | ||
AK085852.1 | - | 3 | 285 | ||
AK142503.1 | - | 3 | 290 | ||
AK158178.1 | - | 3 | 274 | ||
AY007808.1 | - | 3 | 267 | ||
BC031733.1 | - | 3 | 276 | ||
BC058226.1 | - | 3 | 272 | 6308417 |