Gene Transcripts

Gene Symbol
Krtap19-4
Entrez Gene ID
170654
Gene Name
keratin associated protein 19-4
Gene Aliases
Krtap16-4
Chromosome Location
Chr. 16: 88884786 - 88885089 on Build GRCm38
UniGene ID
Mm.196689
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130873.1 NP_570943.1 1 262
GenBank mRNA AF345294.1 - 1 262
BC116200.1 - 1 262 40090087
BC116201.1 - 1 262 40090088

View Full Product Details