Gene Transcripts

Gene Symbol
Nxf7
Entrez Gene ID
170722
Gene Name
nuclear RNA export factor 7
Gene Aliases
Nx7, Nxf6, nfx-a1, nxf-a, nxf-a2
Chromosome Location
Chr. X: 135579555 - 135598777 on Build GRCm38
UniGene ID
Mm.103373
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130888.1 NP_570958.1 10 838
GenBank mRNA AB178901.1 - 11 977
AJ305318.1 - 7 507
AY260550.1 - 10 838

View Full Product Details