Gene Transcripts

Gene Symbol
Muc5ac
Entrez Gene ID
17833
Gene Name
mucin 5, subtypes A and C, tracheobronchial/gastric
Gene Aliases
2210005L13Rik, MGM
Chromosome Location
Chr. 7: 141788958 - 141819217 on Build GRCm38
UniGene ID
Mm.334332
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010844.1 NP_034974.1 50 8125
GenBank mRNA AK008656.1 - 1 167

View Full Product Details