Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008677.2 | NP_032703.2 | 1, 2 | 271 | |
GenBank mRNA | AB002665.1 | - | 1, 2 | 248 | |
AK150510.1 | - | 1, 2 | 253 | ||
AK171315.1 | - | 1, 2 | 49 | ||
BB840090.1 | - | 1, 2 | 283 | ||
BC025517.1 | - | 1, 2 | 100 | 5321182 | |
FJ168470.1 | - | 1, 2 | 62 | ||
U59488.1 | - | 1, 2 | 218 |