Gene Transcripts

Gene Symbol
Nrtn
Entrez Gene ID
18188
Gene Name
neurturin
Gene Aliases
NTN
Chromosome Location
Chr. 17: 56751323 - 56757530 on Build GRCm38
UniGene ID
Mm.8074
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008738.2 NP_032764.1 3 867
GenBank mRNA AK002508.1 - 3 1004
BC057993.1 - 3 848 5345262
U78109.1 - 3 867

View Full Product Details