Gene Transcripts

Gene Symbol
Otog
Entrez Gene ID
18419
Gene Name
otogelin
Gene Aliases
4932420K09, Otgn, twt
Chromosome Location
Chr. 7: 46240987 - 46311434 on Build GRCm38
UniGene ID
Mm.8020
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013624.2 NP_038652.2 16 1767
GenBank mRNA AK165831.1 - 16 1752
U96411.1 - 17 1767

View Full Product Details