Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023418.2 | NP_075907.2 | 2 | 365 | |
GenBank mRNA | AK004921.1 | - | 2 | 221 | |
AK009905.2 | - | 2 | 218 | ||
AK144681.1 | - | 2 | 219 | ||
AK150716.1 | - | 2 | 214 | ||
AK152443.1 | - | 2 | 214 | ||
AK153202.1 | - | 2 | 218 | ||
AK159305.1 | - | 2 | 218 | ||
AK165410.1 | - | 2 | 214 | ||
AK168949.1 | - | 2 | 211 | ||
BC002241.1 | - | 2 | 313 | 3491964 | |
BC005661.1 | - | 2 | 279 | 3588761 | |
BC066844.1 | - | 2 | 202 | 30471755 | |
BC067412.1 | - | 1 | 18 | ||
BC083090.1 | - | 2 | 214 | 30444229 | |
BC106139.1 | - | 2 | 285 | 6528521 |