Gene Transcripts
- Gene Symbol
- Pgam1
- Entrez Gene ID
- 18648
- Gene Name
- phosphoglycerate mutase 1
- Gene Aliases
- 2310050F24Rik, Pgam-1
- Chromosome Location
- Chr. 19: 41911871 - 41918665 on Build GRCm38
- UniGene ID
- Mm.16783
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023418.2 |
NP_075907.2
|
2, 3 |
601 |
|
GenBank mRNA |
AF283667.1 |
- |
2, 3 |
513 |
|
AK004921.1 |
- |
2, 3 |
457 |
|
AK009905.2 |
- |
2, 3 |
454 |
|
AK144681.1 |
- |
2, 3 |
455 |
|
AK150716.1 |
- |
2, 3 |
450 |
|
AK152443.1 |
- |
2, 3 |
450 |
|
AK153202.1 |
- |
2, 3 |
454 |
|
AK159305.1 |
- |
2, 3 |
454 |
|
AK165410.1 |
- |
2, 3 |
450 |
|
AK168949.1 |
- |
2, 3 |
447 |
|
AK185826.1 |
- |
1, 2 |
57 |
|
AK190024.1 |
- |
1, 2 |
57 |
|
AK190271.1 |
- |
1, 2 |
57 |
|
AK192835.1 |
- |
1, 2 |
57 |
|
AK195142.1 |
- |
1, 2 |
57 |
|
AK195515.1 |
- |
1, 2 |
57 |
|
AK196598.1 |
- |
1, 2 |
57 |
|
AK197386.1 |
- |
1, 2 |
57 |
|
AK197934.1 |
- |
1, 2 |
57 |
|
AK199467.1 |
- |
1, 2 |
57 |
|
AK203493.1 |
- |
1, 2 |
57 |
|
AK212756.1 |
- |
1, 2 |
57 |
|
BC002241.1 |
- |
2, 3 |
549 |
3491964 |
BC005661.1 |
- |
2, 3 |
515 |
3588761 |
BC066844.1 |
- |
2, 3 |
438 |
30471755 |
BC067412.1 |
- |
1, 2 |
254 |
|
BC083090.1 |
- |
2, 3 |
450 |
30444229 |
BC106139.1 |
- |
2, 3 |
521 |
6528521 |
View Full Product Details