Gene Transcripts

Gene Symbol
Ptcra
Entrez Gene ID
19208
Gene Name
pre T cell antigen receptor alpha
Gene Aliases
gp33, pT-alpha, pT-alpha-TCR, pT[a], pTa, pTalpha
Chromosome Location
Chr. 17: 46755663 - 46763712 on Build GRCm38
UniGene ID
Mm.215173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011195.2 NP_035325.1 3 511
GenBank mRNA BC119062.1 - 3 436
BC120746.1 - 3 436 40129669
U16958.1 - 3 511

View Full Product Details