Gene Transcripts

Gene Symbol
Lrrc4
Entrez Gene ID
192198
Gene Name
leucine rich repeat containing 4
Gene Aliases
MBAG1, NGL-2, NLG-2, Nag14
Chromosome Location
Chr. 6: 28828126 - 28831747 on Build GRCm38
UniGene ID
Mm.443660
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138682.2 NP_619623.2 1 1576
GenBank mRNA AF300458.1 - 1 1533
AK046781.1 - 1 783
BC012209.1 - 1 1577
BC117833.1 - 1 1517 40092947
BC117834.1 - 1 1517 40092948
DQ177325.1 - 1 1576

View Full Product Details