Gene Transcripts

Gene Symbol
Pthlh
Entrez Gene ID
19227
Gene Name
parathyroid hormone-like peptide
Gene Aliases
PTH-like, Pthrp
Chromosome Location
Chr. 6: 147252110 - 147264085 on Build GRCm38
UniGene ID
Mm.28440
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008970.3 NP_032996.2 3 657
GenBank mRNA AJ278119.1 - 2 152
AY183377.1 - 2 171
AY220497.1 - 3 418
BC058187.1 - 3 462 5346064

View Full Product Details