Gene Transcripts

Gene Symbol
Ptprcap
Entrez Gene ID
19265
Gene Name
protein tyrosine phosphatase, receptor type, C polypeptide-associated protein
Gene Aliases
AI605264, CD-45-AP, LPAP, LSM-1
Chromosome Location
Chr. 19: 4154646 - 4156710 on Build GRCm38
UniGene ID
Mm.271937
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016933.3 NP_058629.2 2 588
GenBank mRNA AI627045.1 - 1 223
AK088334.1 - 2 586
AK165203.1 - 2 586
AW049152.1 - 1 271
BC013273.1 - 2 587 4208992
BC100560.1 - 2 569 832557
D10105.1 - 2 564
U03856.1 - 1 581

Gene Symbol
Rps6kb2
Entrez Gene ID
58988
Gene Name
ribosomal protein S6 kinase, polypeptide 2
Gene Aliases
70kDa, S6K2
Chromosome Location
Chr. 19: 4156792 - 4163383 on Build GRCm38
UniGene ID
Mm.271937
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK090093.1 - 2067

View Full Product Details