Gene Transcripts

Gene Symbol
Purb
Entrez Gene ID
19291
Gene Name
purine rich element binding protein B
Gene Aliases
2310015K15Rik, AA114818, Cager-2, D11Bwg0414e
Chromosome Location
Chr. 11: 6467599 - 6476076 on Build GRCm38
UniGene ID
Mm.296150
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011221.3 NP_035351.1 1 1083
GenBank mRNA AF017630.1 - 1 927
AK049443.1 - 1 220
AK139376.1 - 1 222

View Full Product Details