Gene Transcripts

Gene Symbol
Zzef1
Entrez Gene ID
195018
Gene Name
zinc finger, ZZ-type with EF hand domain 1
Gene Aliases
8430405D05Rik, BC051585, C130099L13Rik, C85424, mKIAA0399
Chromosome Location
Chr. 11: 72796226 - 72927120 on Build GRCm38
UniGene ID
Mm.272030
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045536.2 NP_001039001.1 47 7586
GenBank mRNA AK082066.1 - 1 648
BC099976.1 - 7 942

View Full Product Details