Gene Transcripts

Gene Symbol
Ring1
Entrez Gene ID
19763
Gene Name
ring finger protein 1
Gene Aliases
Ring1A
Chromosome Location
Chr. 17: 34020792 - 34024680 on Build GRCm38
UniGene ID
Mm.20343
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009066.3 NP_033092.3 6 1284
GenBank mRNA AK154962.1 - 6 1245
AK155509.1 - 6 1260
BC009070.1 - 6 1247 2651194
BC082771.1 - 2 2094
BU505852.1 - 1 51
BU918502.1 - 2 327
CT010219.1 - 4 974
CX219100.1 - 1 212
KJ650216.1 - 5 1061
Y12881.1 - 4 980

View Full Product Details