Gene Transcripts

Gene Symbol
Shd
Entrez Gene ID
20420
Gene Name
src homology 2 domain-containing transforming protein D
Gene Aliases
AI413439
Chromosome Location
Chr. 17: 55968392 - 55976625 on Build GRCm38
UniGene ID
Mm.20908
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159523.1 NP_001152995.1 6 997
NM_009168.2 NP_033194.1 6 994
GenBank mRNA AB018423.1 - 6 993
AK142491.1 - 6 1010
AK164639.1 - 6 930
BC052026.1 - 5 1924
BC079574.1 - 5 1924
BC103798.1 - 6 890 30288676

View Full Product Details