Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009258.5 | NP_033284.1 | 3 | 249 | |
GenBank mRNA | AK007841.1 | - | 3 | 210 | |
AK007985.1 | - | 3 | 208 | ||
AV048854.1 | - | 3 | 139 | ||
BC086887.1 | - | 3 | 208 | 6775994 | |
CA481615.1 | - | 3 | 286 | ||
X06342.1 | - | 3 | 206 |