Gene Transcripts

Gene Symbol
Spt1
Entrez Gene ID
20770
Gene Name
salivary protein 1
Gene Aliases
Spt-1
Chromosome Location
Chr. 15: 103895855 - 103899312 on Build GRCm38
UniGene ID
Mm.1280
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC030164.1 - 3 391
BC106151.1 - 3 382 4923478
M33974.1 - 3 394
X00349.1 - 3 395

View Full Product Details