Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172453.3 | NP_766041.1 | 4 | 1525 | |
GenBank mRNA | AK028460.1 | - | 2 | 1713 | |
AK049353.1 | - | 4 | 1527 | ||
AK154072.1 | - | 4 | 1538 | ||
AY100322.1 | - | 5 | 1238 | ||
AY498715.1 | - | 4 | 1525 | ||
BC046611.1 | - | 5 | 1163 | 6396273 | |
BC060108.1 | - | 2 | 1703 |