Gene Transcripts

Gene Symbol
Asb18
Entrez Gene ID
208372
Gene Name
ankyrin repeat and SOCS box-containing 18
Gene Aliases
Asb-18
Chromosome Location
Chr. 1: 89952295 - 90014666 on Build GRCm38
UniGene ID
Mm.215164
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139152.1 NP_631891.1 6 1320
GenBank mRNA AY057054.1 - 6 1320
BC107370.1 - 4 689
BC107371.1 - 5 528
BC127152.1 - 4 689
BC141295.1 - 5 1286 9055907
BC145621.1 - 5 1286 9054189

View Full Product Details