Gene Transcripts

Gene Symbol
Diras1
Entrez Gene ID
208666
Gene Name
DIRAS family, GTP-binding RAS-like 1
Gene Aliases
Di-Ras1, Gbts1, RIG
Chromosome Location
Chr. 10: 81019589 - 81025821 on Build GRCm38
UniGene ID
Mm.44995
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145217.2 NP_660252.1 2 403
GenBank mRNA AK081336.1 - 2 402
AY059640.1 - 1 309
BC057556.1 - 2 403 6810812

View Full Product Details