Gene Transcripts
- Gene Symbol
- Stxbp3
- Entrez Gene ID
- 20912
- Gene Name
- syntaxin binding protein 3
- Gene Aliases
- Munc-18c, Stxbp3a, Sxtbp3, Unc18-3
- Chromosome Location
- Chr. 3: 108793180 - 108840502 on Build GRCm38
- UniGene ID
- Mm.316894
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011504.1 |
NP_035634.1
|
11, 12 |
1002 |
|
GenBank mRNA |
AK138609.1 |
- |
11, 12 |
1034 |
|
AK151466.1 |
- |
11, 12 |
997 |
|
AK152261.1 |
- |
11, 12 |
991 |
|
AK155133.1 |
- |
11, 12 |
1000 |
|
AK169799.1 |
- |
11, 12 |
1003 |
|
BC062901.1 |
- |
11, 12 |
1011 |
6810254 |
BC094055.1 |
- |
11, 12 |
1010 |
|
D30798.1 |
- |
11, 12 |
1002 |
|
U19521.1 |
- |
11, 12 |
1127 |
|
View Full Product Details