Gene Transcripts

Gene Symbol
Wfdc5
Entrez Gene ID
209232
Gene Name
WAP four-disulfide core domain 5
Gene Aliases
BC019734, Prg5, Wap1
Chromosome Location
Chr. 2: 164176325 - 164182742 on Build GRCm38
UniGene ID
Mm.218273
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145369.3 NP_663344.2 2 252
GenBank mRNA AK131905.1 - 2 200
AK131906.1 - 2 185
AY669505.1 - 2 251
BC019734.1 - 2 252 3987617

View Full Product Details