Gene Transcripts

Gene Symbol
Tbc1d4
Entrez Gene ID
210789
Gene Name
TBC1 domain family, member 4
Gene Aliases
5930406J04Rik, A930035N22, AV295684, As160
Chromosome Location
Chr. 14: 101442360 - 101609228 on Build GRCm38
UniGene ID
Mm.320639
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081278.2 NP_001074747.2 1 1192
GenBank mRNA AK031120.1 - 1 1036
AK039337.1 - 1 1193
AK044719.1 - 1 834
BC064433.1 - 1 816
BC080762.1 - 1 810
BC117869.1 - 1 668 40095526
BC117870.1 - 1 668 40095531

View Full Product Details