Gene Transcripts

Gene Symbol
Tdrd5
Entrez Gene ID
214575
Gene Name
tudor domain containing 5
Gene Aliases
Gm103
Chromosome Location
Chr. 1: 156255296 - 156305295 on Build GRCm38
UniGene ID
Mm.252800
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134741.1 NP_001128213.1 8 1172
NM_001277730.1 NP_001264659.1 8 878
GenBank mRNA AK132674.1 - 8 1158
AK133026.1 - 6 543
AY522557.1 - 2 32
BC099972.1 - 8 878 30080156

View Full Product Details