Gene Transcripts

Gene Symbol
Zmat1
Entrez Gene ID
215693
Gene Name
zinc finger, matrin type 1
Gene Aliases
A230096A06, AW990744, B930008K04Rik
Chromosome Location
Chr. X: 134971372 - 135009209 on Build GRCm38
UniGene ID
Mm.336238
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175446.3 NP_780655.2 7 2022
GenBank mRNA AK046974.1 - 2 2152
AK133258.1 - 2019
AK220419.1 - 6 1663
BC067022.1 - 7 2110

View Full Product Details