Gene Transcripts

Gene Symbol
Wdr18
Entrez Gene ID
216156
Gene Name
WD repeat domain 18
Gene Aliases
2310012I10Rik, AU044733, AW122032
Chromosome Location
Chr. 10: 79960152 - 79969246 on Build GRCm38
UniGene ID
Mm.35828
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175450.4 NP_780659.2 3 385
GenBank mRNA AK050106.1 - 3 383
AK082695.1 - 3 387
AK133716.1 - 3 386
AK160713.1 - 3 383
AK166113.1 - 383
AK168691.1 - 3 383
BC032968.1 - 2 77
BC095984.1 - 3 385 4978914

View Full Product Details